Would you like to log in again?
AlphaTec® 88-390T Neoprene and Natural Rubber Latex Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AlphaTec®
193951 193953 193955
89089-172CS 471.83 USD InStock
89089-172 89089-170 32889-654
AlphaTec® 88-390T Neoprene and Natural Rubber Latex Gloves, Ansell
Gloves
Type 88-390 gloves provide the right mix of chemical protection, dexterity and sensitivity for mid-level applications.
  • Excellent chemical resistance
  • Pebble embossed design increases grip on wet or dry surfaces
  • Durable
The 13-millimeter blend of neoprene and natural rubber latex delivers optimal durability and chemical resistance.
Use in the aerospace, automotive and transportation, chemical, food processing, food service, petrochemical, pharmaceuticals, and general industries. Uses include canning and food processing; detergents and cleaning liquids; handling acids, alkalies, salts, ketones and alcohols; and janitorial and maintenance work.
Certifications: Complies with FDA food handling requirements (21 CFR 177.2600).
Ordering information: This product contains natural rubber latex, which may cause allergic reactions in some individuals.
Packaging: Gloves are packaged 12 pairs in a bag; 144 in a carton.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty