Would you like to log in again?
AlphaTec® Solvex® Nitrile gloves, Ansell
Supplier: Ansell
AlphaTec® Solvex®
117274 3716511090 117074 3715511090 3714511090 117141 3717511100 3716511100 3715511110 3715511070 3715511110 3716511080 3714511080 3714511070 3715511100 3715511080 117076 117209 3714511100 3717511070 3717511080 3718511100 117272 117077 3714511110 3718511090 117210 3715511090 3715511100 117275 117073 117276 117208 117211 117075 3716511110 3717511090 117142 3717511110 117143 117071 3716511070 3715511080 117144 117273 117145 117277 117207
32890-124CS 1363.64 USD InStock
32890-124 32890-122 CA32890-116 32890-088 CA32890-118 32890-120 CA32490-009 CA32490-008 32890-086 CA32890-094 CA32890-096 32890-108 32890-106 32890-128 32890-126 CA-32890-130 CA32890-090 CA-32890-132 CA32890-092 CA-32890-134 CA-32890-136 CA32890-124 32890-114 32890-136 32890-134 32890-112 32890-132 32890-110 32890-130 32890-096 CA32490-015 CA32890-140 CA32890-120 CA32890-142 32890-118 CA32890-088 CA32890-122 CA32490-011 32890-116 CA32490-010 CA10798-912 CA10798-934 CA10798-910 CA10798-932 32890-094 CA-32890-128 32890-092 32890-090
AlphaTec® Solvex® Nitrile gloves, Ansell
Gloves
Chemical-resistant gloves are suitable for chemical processing, food processing, aerospace and automotive degreasing, general manufacturing, electronics, and other applications.
  • Curved finger design and contoured palm to reduce fatigue
  • Embossed texture improves grip for easier handling of wet objects
  • Hand-specific
  • Powder-free
  • Straight cuffs
Tough nitrile compound provides long wear and good protection against abrasion, cuts, snags, and punctures. Strong resistance to degradation ensures that gloves will not weaken or swell. Optional cotton flock lining absorbs perspiration and makes donning and doffing easier. Gloves are free of protein allergens and will not promote contact dermatitis.
Protects against a wide range of chemicals, including aromatic solvents, cutting oils and coolants, petroleum solvents and animal fats.
Certifications: Component materials comply with FDA food handling requirements (21 CFR 177.2600).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty