Would you like to log in again?
Qualatex® XC 7J Anti-Static Latex Fingercots, QRP
Supplier: QRP
7JM 7JXL 7JL 7JS
32891-154CS 1167.4 USD InStock
32891-154 32891-150 32891-152 32891-148
Qualatex® XC 7J Anti-Static Latex Fingercots, QRP
Finger Cots Latex Finger Cots
The QRP 7J Anti-Static Clean Room Latex Finger Cots are recommended for use with static sensitive devices.
  • Packaged for ISO 5 (Class 100) applications
  • Compliant with ASTM and IES-RP-CC 005.3 standards
  • Average surface resistance of 5 X 10¹² ohms per square unit
  • For use with Class II static sensitive devices (thresholds above 1000V)
7C pink Anti-Static Clean Room Latex Finger Cots are recommended for use with static sensitive devices. The QRP 7C gloves are packaged for ISO 5 (Class 100) applications and are compliant with ASTM and IES-RP-CC 005.3 standards. Finger cots are packaged in gas-impremeable film flushed with semiconductor grade N2.
Certifications: ESD Safe to ANSI/ESD S20.20-2014.
Caution: This product contains natural rubber latex which may cause allergic reactions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty