Would you like to log in again?
Vinyl Coat Aprons, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
950192 950191 950193 950194 950190
32890-884CS 458.65 USD InStock
32890-884 32891-286 32890-882 32890-889 32890-888
Vinyl Coat Aprons, Ansell
Disposable Garments
SType 56-910 aprons provide excellent protection when handling acids and caustic solutions.
  • Heat-sealed binding at the neck and bottom eliminates leaks caused by exposed elastic and thread
  • Back closure consists of two plastic snaps and nylon ties, with no metal parts to cause static or corrosion
  • Full-length sleeves provide additional coverage
Made of virgin vinyl.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty