Would you like to log in again?
Magid® Heater Beater® 499KBT Cotton Canvas Hot Mill Gloves, Magid
Supplier: Magid
499KBT
32893-117PK 115.26 USD InStock
32893-117
Magid® Heater Beater® 499KBT Cotton Canvas Hot Mill Gloves, Magid
Gloves
Magid® Heater Beater® Hot Mill Gloves keep out the heat and provide superior comfort and protection.
  • 31 oz. weight keeps out the heat
  • Heavy-duty triple-layer, nap out construction
  • Knuckle strap for added protection
  • Pull patch for increased durability
Unique triple layer design features heat-resistant burlap between outer and inner layers of cotton for maximum heat protection and comfort.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty