Would you like to log in again?
Magid® RoadMaster® Leather Gloves, Magid
Supplier: Magid
BU500-S BU500-XL
32893-124PK 159.7 USD InStock
32893-124 32893-126
Magid® RoadMaster® Leather Gloves, Magid
Gloves
Top grain deerskin leather gloves offers superior flexibility, softness and durability.
  • Gunn cut pattern delivers optimal dexterity
  • Shirred elastic at wrist holds gloves securely on the user's hand
  • Keystone thumb with reinforced stitching provides maximum comfort, grip and wear
This dependable deerskin grain drivers glove also features a gunn cut pattern and a keystone thumb for optimal dexterity and maximum comfort.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty