Would you like to log in again?
Magid® Top Gunn® Select Double Leather Palm Cuff, Magid
Supplier: Magid
TB257EDP
32893-169PK 79.8 USD InStock
32893-169
Magid® Top Gunn® Select Double Leather Palm Cuff, Magid
Gloves
Magid® Top Gunn cow split leather palm gloves have a double layer of leather over the palm area for reduced wear and tear and added grip. These sturdy gloves feature a wing thumb, knuckle strap, pull patch, lined palm and elastic for comfort and easy use.
  • Elastic back improves comfort and fit
  • Double layer of leather over palm, index and thumb
The glove recommended for rugged jobs that require extra protection for the back of the wearer's hand and fingers. This durable style also features a 2¹/₂" safety cuff that extends protection beyond the wrists.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty