Would you like to log in again?
GLOVES 5MIL VINYL YEL LRG PK24
Supplier: NEWELL BRANDS INC
PYLOX* Vinyl Gloves^ Large. Length: 26.7cm. Color: yellow. 5mil thickness. Unlined, pinked cuff. Hand-specific. Light bisque finish provides safe grip for wet or dry applications. Lightly powdered for easy on-off. Liquid-tight. Nitrile in acids, caustics, alcohols, aldehydes, oils, and grease.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES 5MIL VINYL YEL LRG PK24 32916-016 541968 Case of 288 Retrieving
Pack of 24 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty