Would you like to log in again?
GLOVES TR.PLY TRI 14IN 20MIL SZ7 PK12
Supplier: NEWELL BRANDS INC
TRIonic* E-194BPK Controlled Environment Acid Gloves^ Natural rubber latex, neoprene, and nitrile blend for chemical resistance. Meet CFR 21 170-199 for food and drug handling. Color: Natural. Embossed "Z" grip. 20mil thickness. 35.6cm length. Size: 7. Bulk packed.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES TR.PLY TRI 14IN 20MIL SZ7 PK12 32916-102 517987 Case of 72 Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty