Would you like to log in again?
GLOVES NITRL MDWT GRN SZ7 PK12
Supplier: NEWELL BRANDS INC
STAN-SOLV* Nitrile Gloves^ Size: 7. Length: 35.5cm. Green. Mediumweight. Knit-lined. Separate shell-and-liner construction prevents leaks paths. Soft, two-way-stretch interlock cotton liner absorbs perspiration and provides insulation. Z-pattern grip channels fluids away for secure grip.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES NITRL MDWT GRN SZ7 PK12 32916-178 34381047 Case of 72 Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty