Would you like to log in again?
TRIonic® Tri-Polymer Gloves, Mapa Professional AdvanTech
Supplier: Mapa Professionnel
TRIonic®
521319 521318 514316 514318 521310 514319 514310 514311 514317
32930-798CS 411.76 USD InStock
32930-798 32930-788 32916-352 32930-791 32930-792 32916-350 32930-794 32930-796 32916-354
TRIonic® Tri-Polymer Gloves, Mapa Professional AdvanTech
Gloves
Unique tri-polymer construction offers 100% comfort for optimal mechanical and chemical resistance. Tri-polymer blend of neoprene, nitrile, and natural rubber combines comfort with added strength, tear resistance, and chemical splash protection.
  • Low level of contaminants
  • Extensive range for all applications
  • Wide cuff for double-gloving
  • Extended length to protect the forearm
This product is not classified as hazardous according to the directive 1999/45/EC of the European Parliament and of the Council.
This product does not contain any substances of very high concern according to the regulation n° 1907/2006 of the European Parliament and of the Council (REACH).
Certifications: Compatible with ISO 4 class 10.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty