Would you like to log in again?
Evolution One™ Latex Gloves, Microflex®, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Evolution One™
EV-2050-L EV-2050-XL EV-2050-M EV-2050-XS EV-2050-S EV-2050-S EV-2050-XS EV-2050-M EV-2050-L EV-2050-XL
32916-536CS 339.29 USD InStock
CA32916-536 32916-536 CA32916-538 32916-534 CA32916-532 CA32916-534 32916-538 CA32916-530 32916-532 32916-530
Evolution One™ Latex Gloves, Microflex®, Ansell
Gloves
These ambidextrous latex gloves have added coating that provides super antisticking properties.
  • Ambidextrous
  • Interior polymer coating for superior antisticking properties in all conditions
  • Fully textured, beaded cuff, powder-free
  • Thickness: 8.7 mil (finger)
These gloves have an interior polymer coating that provides superior antisticking properties, resulting in highly effective donning in wet, damp, or dry conditions.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty