Would you like to log in again?
Supreno® EC Powder-Free Nitrile Gloves, Microflex®, Ansell
Supplier: Ansell
Supreno®
SEC-375-XS SEC-375-L SEC-375-L SEC-375-S SEC-375-XS SEC-375-XXL SEC-375-XXL SEC-375-S SEC-375-M SEC-375-3XL SEC-375-M SEC-375-XL SEC-375-XL SEC-375-XS SEC-375-3XL
32916-678CS 293.82 USD InStock
32916-678 89135-222 CA32916-676 CA10755-079 CA10755-078 CA32916-678 32916-672 76311-656 CA32916-672 32916-670 CA32916-674 32916-676 32916-674 CA10755-082 CA32916-670
Supreno® EC Powder-Free Nitrile Gloves, Microflex®, Ansell
Gloves
Soft, durable nitrile formulation delivers improved comfort, strength and durability for maximum protection against rips, snags or tears
  • Ambidextrous
  • Powder-free
  • Beaded cuffs
  • Online chlorination
Gloves are suitable for high-risk applications and feature an extended cuff to protect the wrist and forearm. Interior polymer coating makes gloves easy to don. Textured fingers allow for reliable grip in wet and dry conditions.
Designed for workers in high risk environments, this glove features a low 0.65 AQL for advanced barrier protection.
Certifications: Gloves meet or exceed the emergency medical glove requirements of NFPA 1999. Tested for use with chemotherapy/cytotoxic drugs.
Packaging: Gloves are packaged 50 gloves per dispenser box.
Caution: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty