Would you like to log in again?
CE4 System Sterile Latex Hand-Specific Gloves, Microflex®, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HSCE4-879-85 HSCE4-879-80 HSCE4-879-60 HSCE4-879-70 HSCE4-879-65 HSCE4-879-90 HSCE4-879-75
32916-746CS 866.18 USD InStock
32916-746 32916-744 32916-748 32916-742 32916-752 32916-740 32916-750
CE4 System Sterile Latex Hand-Specific Gloves, Microflex®, Ansell
Gloves
These hand specific gloves are packaged with a double vacuum seal for use in ISO Class 4 (FED-STD-209E Class 10/M2.5) controlled environments.
  • Silicone-free
  • Micro-textured surface enhances wet or dry grip
  • Provides superb aesthetics, dexterity, tactile sense, fit, feel, and comfort
  • Powder-free
These gloves are washed in microprocessor-controlled, two-in-one, washer/dryer laundering systems to provide very low surface particulates, ionic extractables, and non-volatile residues.
Certifications: Lot-specific Certificate of Analysis available upon request.
Packaging: Pair-packed in individual wallets, 10 wallets per vacuum-sealed poly bag, 20 bags per case.
Caution: These gloves contain natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty