Would you like to log in again?
Microflex® FreeForm® EC FFE-775 Examination Gloves, Ansell Healthcare, Ansell
Supplier: Ansell
CR100® FreeForm®
FFE-775-XXL FFE-775-M FFE-775-L FFE-775-XL FFE-775-S FFE-775-L FFE-775-XL FFE-775-XXL FFE-775-M FFE-775-S
32916-778CS 302.38 USD InStock
CA32916-778 32916-778 CA32916-786 32916-782 CA32916-782 32916-780 CA32916-784 32916-786 32916-784 CA32916-780
Microflex® FreeForm® EC FFE-775 Examination Gloves, Ansell Healthcare, Ansell
Gloves
Extended protection and grip, approved for use by first responders.
  • Non-stick properties resists tape and adhesives
When performing mild-duty work, MICROFLEX® FreeForm® EC is a long cuff nitrile glove that offers reliable protection and a secure grip. Its extended cuff provides expanded protection over the wrist and forearm.
Certifications: ASTM D6319, EN 420:2003 + A1:2009, FDA21 CFR 177-2600, NFPA 2012, US QSR/FDA 510(k) Medical Examination Grade
Packaging: Gloves are packaged 50 per dispenser, 10 dispensers per case (500 gloves per case).
Caution: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty