Would you like to log in again?
Technic™ Flock-Lined Neoprene-Over-Natural-Rubber Gloves,
Supplier: Mapa Professionnel
Technic™
420440 401448 450440 420448 401447 401449 420446 420449 450449 450447 450448 401446 420447 401440
32917-239CS 209.85 USD InStock
32917-239 32917-229 32917-240 32917-242 32917-231 32917-220 32917-241 32917-230 32917-233 32917-222 32917-221 32917-232 32917-224 32917-223
Technic™ Flock-Lined Neoprene-Over-Natural-Rubber Gloves,
Gloves
Gloves offer reliable hand protection for chemical processing, metal finishing, mechanical work, painting, degreasing, plating, bleaching, leather tanning, commercial dish/pot washing, janitorial, and general maintenance applications.
  • Color: black
  • Comfortable flock lining
  • Gloves are chemical-resistant and heavy-duty
Flexible, comfortable, hand-specific design features a flat cuff and curved fingers for a natural fit. Embossed diamond grip for safe, easy handling of wet and greasy objects.
Warning: This product contains natural rubber latex and is not recommended for those who are allergic to natural rubber proteins.
Certifications: In compliance with 21 CFR 170-199. CE marked.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty