Would you like to log in again?
GLOVES 30MIL NEOPRN BLK SZ9 PK
Supplier: NEWELL BRANDS INC
CHEM-PLY* Neoprene Gloves^ Size: 9. Length: 46cm. Color: black. 30mil thickness. Unlined style. Curved-finger, contoured-palm design for fit and comfort. Embossed Z-pattern grip channels fluids away from working surface to provide positive non-slip grip on hard-to-grasp materials.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES 30MIL NEOPRN BLK SZ9 PK 32917-274 414709 Case of 12 Retrieving
Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty