Would you like to log in again?
STANSOLV® Flock-Lined Nitrile Gloves, Mapa Professionnel
Supplier: Mapa Professionnel
STANSOLV®
483427ZQK 493429 483429ZQK 493428 483428ZQK 483420ZQK 483421 483426 493420
32917-700CS 312.09 USD InStock
32917-700 32917-707 32917-729 32917-718 32930-110 32930-111 32930-112 32917-730 32917-740
STANSOLV® Flock-Lined Nitrile Gloves, Mapa Professionnel
Gloves
Offer hand protection for industrial applications such as plating, metal finishing, chemical processing, abrasive material handling, maintenance, and acid etching.
  • Curved fingers and tapered wrist for natural, comfortable wear
  • Easy donning and doffing
  • Flock lining absorbs perspiration for added comfort
  • Flat cuffs
  • Hand-specific
Double-dipped gloves are highly resistant to aromatic petroleum and chlorinated solvents, acids, alkalies, grease, oils, and Freon®. Nitrile formulation is free of inert fillers and silicone for superior chemical resistance. Gloves contain no natural rubber proteins and will not cause allergic reactions. Also resist punctures, snags, cuts, and abrasions.
Color: Green
Certifications: In compliance with 21 CFR 170-199. CE marked.
Caution: Note: Not recommended for use with ketones, strong oxidizing acids, and organic chemicals containing nitrogen.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty