Would you like to log in again?
STANSOLV® Nitrile Gloves, AdvanTech, Mapa Professionnel
Supplier: Mapa Professionnel
STANSOLV®
519310 519316 519318 519317 519311 519319
32917-855CS 529.88 USD InStock
32917-855 32917-857 32917-856 32917-859 32917-858 32917-860
STANSOLV® Nitrile Gloves, AdvanTech, Mapa Professionnel
Gloves
STANSOLV® A-10CR gloves provide hand protection against solvents and other chemicals that attack natural rubber. Ideal for solvent-based photo-resists, caustic developer solutions, and organic-based strippers. Used for semiconductor wet processes using solvent-based strippers, etchants, and cleaners. Also used for handling cleanroom and clean process acids, caustics, and solvents.
  • Case-hardened finish for added chemical resistance
  • Easy on/off
Gloves resist organic solvents such as xylene, perchloroethylene, freon, ethylene glycol ether, and Cellosolve®
Hand-specific gloves are powder-free with flat cuffs.
Cut-, puncture-, and abrasion-resistant. Unique embossed Z-pattern grip improves wet gripping. Finished, tested, and packaged in Class 100 cleanroom and 100% inspected for leaks. Gloves contain no natural rubber proteins and will not cause allergic reactions.
Certifications: In compliance with 21 CFR 170-199. CE approved. Class M 2.5.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty