Would you like to log in again?
GLOVES,A-18,NITRILE CS-12PR 11
Supplier: NEWELL BRANDS INC
STANSOLV* Nitrile Gloves^ A-18. Green. Hand-specific. Case-hardened finish. Resist most solvents, oil, grease, fats, acids, alkalis, alcohols. Resist punctures, abrasion. Curved finger. Tapered wrist. Flat cuff. Comply with 21 CFR 170-199. Embossed Z-pattern grip. 22mil thick. 45.7cm long. Size: 11.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES,A-18,NITRILE CS-12PR 11 32929-885 480431 Case of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty