Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Blue-Grip™ Natural Rubber Gloves, Mapa Professionnel

Supplier: Mapa Professionnel
Blue-Grip™
301426 301427 301428 301429
32929-901CS 458.45 USD InStock
32929-901 32929-902 32929-900 32929-903
Blue-Grip™ Natural Rubber Gloves, Mapa Professionnel
Gloves
MAPA LL-301 gloves offer hand protection for such applications as food processing, handling frozen or hot containers, agricultural chemicals, masonry, sheet metal assembly, heavy-duty cleaning, and maintenance.

  • Pinked cuff
  • Color: light blue/medium blue
  • Washable without ball-up of cotton
  • Heavy-gauge natural latex rubber gives excellent mechanical and chemical protection
  • Latex coating is firmly bonded to 255 g (9 oz.) cotton knit lining

Double-dipped work area has etched nonscratch, nonabrasive, nonslip grip. Curved contour anatomical design; hand-specific. Lining absorbs perspiration, adds insulation and comfort, and provides easy donning and doffing. Single-seam lining with no seams on work surface. Heavy-duty flexible liquid-proof gauntlet gloves resist acids, caustics, alcohol, detergents, germicides, salts, aldehydes, and ketonic solutions, as well as abrasions, tears, cuts, and punctures.

Certifications: In compliance with CFR 21 170-199. CE marked.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty