Would you like to log in again?
Classics Natural Rubber Gloves, Mapa Professional
Supplier: Mapa Professionnel
177017
32930-671CS 279.07 USD InStock
32930-671
Classics Natural Rubber Gloves, Mapa Professional
Gloves
These natural rubber gloves are designed for genenral lab applications.
  • Chlorinated interior for easy on-off
  • Embossed diamond grip pattern
  • Unlined
Grip design prevents build up from greasy solids. The user can easily maintain a positive grip even on hard-to-grasp materials.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty