Would you like to log in again?
GLOVES SOLO ULTRA L PK100
Supplier: NEWELL BRANDS INC
Powder Free Nitrile Gloves^ Solo Ultra* 997 disposable 100% powder free ambidextrous nitrile exam gloves. Meets ASTM F1671 resistance to blood borne pathogens. Textured fingertips provide safe wet or dry grip. Rolled cuff. Blue. 4mil. 25 cm length. Size: Large
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES SOLO ULTRA L PK100 32930-744 34977168 Case of 1000 Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty