Would you like to log in again?
Solo Ultra 980 Nitrile Gloves, Mapa Professional®
Supplier: Mapa Professionnel
980428 980426 980421 980420 980427
32930-776CS 985.7 USD InStock
32930-776 32930-778 32930-780 32930-784 32930-786
Solo Ultra 980 Nitrile Gloves, Mapa Professional®
Gloves
Nitrile gloves with excellent mechanical resistance, ideal in oily environments and manufacturing of medicines
  • Textured grip offers added safety when working in wet and dry conditions
  • Exterior finish: smooth with pebbled fingertips
  • Chlorinated interior finish
Packaging: 100 gloves/box and 1000 gloves/carton.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty