Would you like to log in again?
GLOVE TRIPOLY 514+ SZ7 PK12
Supplier: NEWELL BRANDS INC
TRIonic* 514 Plus Gloves^ Feature unique tri-polymer blend of natural latex, nitrile, neoprene with diamond grip. Hand-specific, powder-free. Produced in Class 100 cleanrooms. Meet CFR 21 170.199. 20 mil thickness. 38 cm length. Natural. Size: 7. Pair packed.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVE TRIPOLY 514+ SZ7 PK12 32930-790 514317 Case of 72 (1/ Itm/Bx) Retrieving
Pack of 12 (1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty