Would you like to log in again?
STANZOIL® Neoprene-Coated Knit-Lined Gloves, Mapa Professionnel
Supplier: Mapa Professionnel
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
STANZOIL®
334949 337429 337428 334946 334947 337420 337421 334948
32920-086CS 942.5 USD InStock
32920-086 32920-020 32920-042 32920-064 32931-490 32931-515 32931-537 32931-559
STANZOIL® Neoprene-Coated Knit-Lined Gloves, Mapa Professionnel
Gloves
Gloves provide medium-weight hand protection for chemical processing, petroleum refining, grinding and filling of castings, metal finishing, and general maintenance applications.
  • Soft, absorbent interlock knit-cotton liner
  • Washable
  • Mechanical and chemical protection
Liquid-proof and resistant to oils, acids, caustics, solvents, greases, abrasion, and cuts. adds comfort, easy on-off, and moderate temperature protection. Roughed overdip provides a nonslip grip for hard-to-grasp materials. Two-color construction for a visual indicator of wear-through.
Each style features a unique multi-layer construction, avoiding the thin spots and pinholes common to single-dipped supported gloves. Durable neoprene for excellent dexterity.
Certifications: In compliance with CFR 21 170-199. CE marked.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty