Would you like to log in again?
Rollpruf® Cleanroom Gloves, AdvanTech, Mapa Professionnel
Supplier: Mapa Professionnel
Rollpruf®
513564 513987 513985 513984 513986 513566 513567 513565
32931-730CS 847.34 USD InStock
32931-730 32931-774 32931-796 32931-752 32932-132 32932-110 32932-096 32932-074
Rollpruf® Cleanroom Gloves, AdvanTech, Mapa Professionnel
Gloves
ISO Class 4 (FED-STD-209E Class 10/M2.5) gloves provide product protection for hand tasks in cleanroom applications including assembly, testing, and inspection in the semiconductor, optical, aerospace, and electronics industries.
  • Triple-polymer blend of natural rubber latex, nitrile, and neoprene
  • Hand-specific
Eextractables, and nonvolatile residue (NVR). Color-coded cuff band prevents roll down. Powder-free. Textured bisque grip. Color: natural with color-coded cuff band.
Certifications: Meet CFR 21 170-199 for food and drug handling, and IEST-RPCC-0052 wet tests for particulate, total extractables, and contribution to corrosion.
Ordering information: Individual pairs packaged in sealed antistatic bags and supplied in two antistatic master poly bags of 100 pair each. Bulk packaging consists of four sealed poly bags, 50 singles each.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty