Would you like to log in again?
Latex Gloves, Hand-Specific, Kimtech™ G3, Kimberly-Clark
Supplier: Kimberly-Clark
Kimtech™ G3
56825 56833 56835 56829 56831 56838 56839 56836 HC1385 56831 56827 HC1360 56836 56841 56828 56832 56830 56840 56834 HC1370 56830 56825 56841 56837 HC1310 56833 56832 56827
32933-728CS 553.35 USD InStock
CA32933-716 CA32933-738 32933-728 CA32933-736 CA32933-813 32933-726 CA32933-718 CA32933-730 32933-742 32933-740 CA32933-734 CA32933-811 32933-724 32933-722 32933-718 CA32933-726 32933-716 32933-738 CA32933-724 CA32933-728 32933-732 CA32933-740 32933-736 32933-813 32933-734 32933-811 CA32933-742 CA32933-720
Latex Gloves, Hand-Specific, Kimtech™ G3, Kimberly-Clark
Gloves
Designed for critical cleanroom environments.
  • Double-bagged to ensure cleanliness
  • Textured palms and fingertips
Gloves offer exceptional wet/dry grip without compromising tactile sensitivity. The tapered, beaded cuff reduces rolldown to make donning easier, enhances cuff strength, and forms a better seal on the garment. AQL 1.5.
Specially processed gloves are washed multiple times in ultrapure deionized water to ensure consistent control of particles and extractables.
Certifications: Recommended for ISO Class 4 or higher cleanroom environments.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established. This product should not be worn by or exposed to individuals allergic to natural rubber latex.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty