Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PURPLE NITRILE™ and PURPLE NITRILE-XTRA™ Examination Gloves, Kimberly-Clark

Supplier: Kimberly-Clark
KIMBERLY-CLARK® PURPLE NITRILE®
55090 50603 50601 55083 55082 50604 55081 55080 55082 55084 55090 50603 50602 50601 50602 55080 50604 55084 55081 55083
32934-082CS 428.11 USD InStock
CA32933-968 32934-082 CA32934-080 32933-968 32934-084 CA32934-082 32934-076 CA32934-084 32934-078 CA32934-076 32933-974 CA32933-972 CA32933-970 32933-972 CA32933-976 CA32933-974 32933-976 32934-080 32933-970 CA32934-078
PURPLE NITRILE™ and PURPLE NITRILE-XTRA™ Examination Gloves, Kimberly-Clark
Gloves
Nitrile gloves offer excellent tactile sensitivity and dexterity as well as superior fit and comfort.

  • Offer excellent tactile sensitivity and dexterity
  • Textured fingertips and beaded cuff
  • Ambidextrous and powder-free
  • Length: 24.1 cm (9.5") or 30.5 cm (12")
  • Thickness: 5.9 mil (finger), 4.7 mil (palm), 3.5 mil (cuff)

Beaded cuff for added strength and easier donning. Glove fingers are thicker for greater barrier protection. Textured fingertips enhance wet and dry grip. Ambidextrous and powder-free. Color: purple.

Certifications: Exceed ASTM standard for critical defects. Meets Chemotherapy Permeation Testing per ASTM D6978-05.

Ordering information: Packaged in dispenser boxes.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty