Would you like to log in again?
Spectra® Fiber Cut-Resistant Glove Liners, Wells Lamont
Supplier: Wells Lamont
Spectra®
M214L M214M M214S M214XS M214XL
32935-487BG 402.17 USD InStock
32935-487 32935-485 32935-483 32935-509 32935-481
Spectra® Fiber Cut-Resistant Glove Liners, Wells Lamont
Gloves
An ultrahigh molecular weight polyethylene fiber provides moderate cut resistance with high reusability.
  • Gloves can be used outside
Gloves are lightweight and have a low coefficient of friction, which allows sharp objects to slide across the surface.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty