Would you like to log in again?
GLOVE LINER NYL GRN SM BG=20PR
Supplier: WELLS LAMONT INDUSTRY GROUP
Nylon Glove Liner^ Reusable, thin, nylon glove liner provides a protective barrier between user's hands and latex gloves to prevent skin irritation without affecting tactile sensitivity. Ambidextrous, full-finger style with regular cuff. Size: Small.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVE LINER NYL GRN SM BG=20PR 32935-580 M555S Bag of 20 Retrieving
Case of 1200 Retrieving
Pack of 200 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty