Would you like to log in again?
EYEGLASSES CLEAR LENS 78008
Supplier: Avantor
Safeguards* Eye Glasses^ Designed for comfort and protection from eye damage caused by flying particles and dust. Clear lens. Charcoal frame. Meet ANSI Z87.1-1989 standard. SEI certified.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EYEGLASSES CLEAR LENS 78008 33000-125   Each Retrieving
Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty