Would you like to log in again?
EYEWEAR SEBRING400 PC BLK/CLR
Supplier: SUREWERX USA INC
Sebring 400* Safety Glasses^ One-piece, integral lens. Lightweight, plastic frame. Hard-coated polycarbonate lens. Temple length: 14.6cm Meets ANSI Z87.1-1989 Standard. Black frame with clear lens.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EYEWEAR SEBRING400 PC BLK/CLR 33000-245 S76301 Each Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty