Would you like to log in again?
Uvex Stealth® Goggles, Honeywell Safety
Supplier: Honeywell Safety
uvex stealth®
S3960HS S3961C S3960D S3960C S3970DF S39611C S39611C S3960CI S3960D S3961C S3970D S3971D S39610C S3961HS S3970D S3960CI S3962C S3962C
33000-805CS 1192.53 USD InStock
CA33000-805 33000-805 33000-756 33000-755 10790-662 CA33000-752 CA89095-448 10790-630 CA33000-756 CA94023-954 89095-456 94023-954 33000-752 33000-751 CA89095-446 CA33000-760 89095-446 33000-760
Uvex Stealth® Goggles, Honeywell Safety
Goggles
Protect against dust, mist, and splashes with these stylish goggles.
  • Durable polypropylene frame and soft, flexible elastomer provide comfortable seal
  • Indirect ventilation reduces fogging
  • Uvextreme® lenses provide antifog, anti-fog, anti-scratch, anti-static
  • Dura-streme® lenses feature interior Uvextreme® coating and exterior Ultra-dura® coating for enhanced durability
  • One-piece, impact-resistant polycarbonate lens can be replaced quickly and easily
Lenses are available with either the exclusive Uvextreme® AF or Dura-streme™ lens coatings. Dura-streme™ lens coating features a combination of Ultra-dura® on the exterior and Uvextreme® on the interior for excellent protection and longevity. The Rx Carrier goggle insert is an accessory item designed to hold prescription lenses in place inside the goggle.
Durable polypropylene frame, combined with soft, flexible elastomer, conforms to facial contours for a comfortable seal.
Certifications: Meet ANSI Z87+ and CSA Z94.3 high impact protector standards.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty