Would you like to log in again?
Centurion™ 452 Series Impact Safety Goggles, 3M
Supplier: 3M
Centurion™
7100009575 7100009575
33001-599CS 214.13 USD InStock
33001-599 CA33001-599
Centurion™ 452 Series Impact Safety Goggles, 3M
Goggles
Centurion™ Safety Impact Goggle is a reliable choice in protective eyewear. Stable, sturdy construction will help protect wearer's eyes.
  • Wide-angle wraparound lens and frame for good peripheral vision
  • Direct venting system to help minimize fogging
  • Accommodates most prescription eyewear
Includes wide nasal flare for added comfort and a direct venting system to minimize fogging. All goggles feature a strap attachment system.
Certifications: Meets ANSI Z87.1-2010, CSA Z94.3-2007.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty