Would you like to log in again?
Centurion™ 454 Series Splash Safety Goggles, 3M™ Company
Supplier: 3M
Centurion™
7000002377 7000127565 7000002377
33001-601CS 197.82 USD InStock
33001-601 33001-603 CA33001-603
Centurion™ 454 Series Splash Safety Goggles, 3M™ Company
Goggles
Centurion™ safety splash goggle is a reliable, economical choice in protective eyewear. Stable, sturdy construction will protect wearer's eyes.
  • Wide-angle wraparound lens and frame for good peripheral vision
  • Indirect venting system to help minimize fogging
  • Accommodates most prescription eyewear
Includes wide nasal flare for added comfort and an indirect venting system to minimize fogging. All goggles feature a strap attachment system.
Certifications: Meets ANSI Z87.1-2010, CSA Z94.3-2007.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty