Would you like to log in again?
KIMAX® Pharmaceutical Graduated Cylinders, Double Scale, Conical, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMAX®
60345-4 60345-2 60345-80 60345-32 60345-120 60345-16 60345-60 60345-8 60345-40
33028-194CS 921.05 USD InStock
33028-194 33032-126 33032-104 33028-193 33032-148 33028-191 33032-181 33028-197 89002-238
KIMAX® Pharmaceutical Graduated Cylinders, Double Scale, Conical, Kimble Chase
Cylinders Measuring Cylinders
Manufactured from borosilicate glass 3.3. Each instrument is individually calibrated and certified against equipment whose calibration is traceable to the National Institute of Standards and Technology.
  • ''TD'' appears on each cylinder and indicates the cylinder is calibrated to deliver
  • Double scale features both metric and U.S. customary units
  • Scale is durable white ceramic enamel
Tolerances for the various lines on the graduates depend on the inside diameters of these lines, and may be found in the handbook. Designed from ASTM specification E1094, Type 3 requirements.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty