Would you like to log in again?
Separatory Funnel with Detachable TFE Drip Tips, Chemglass
Supplier: Chemglass
CG-1744-01 CG-1744-26 CG-1744-33 CG-1744-15 CG-1744-03 CG-1744-25 CG-1744-43 CG-1744-04 CG-1744-05 CG-1744-32 CG-1744-17 CG-1744-34 CG-1744-18 CG-1744-23 CG-1744-02 CG-1744-30 CG-1744-16 CG-1744-24 CG-1744-42 CG-1744-31
89028-658EA 219.79 USD InStock
89028-658 89028-668 33500-278 33500-286 33500-276 89028-670 89028-674 33500-280 33500-270 89028-684 89028-662 89028-672 89028-660 89028-682 33500-284 33500-274 89028-676 33500-282 89028-664 33500-272
Separatory Funnel with Detachable TFE Drip Tips, Chemglass
Funnels Separating Funnels
Squibb funnel with a PFTE stopcock. The TFE stem attaches directly to the glass stopcock with O-Rings or other gaskets.
  • Detachable G-14 or G-18 TFE drip tip
  • Supplied with PE or glass stopper of the listed size
This unique separatory funnel is manufactured with a removable PTFE stem that virtually eliminates breakage normally associated with standard glass stem stopcocks. Glassware equipped with this PTFE stem can survive repeated forceful strikes to the stem without breakage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty