Would you like to log in again?
PROSAT® Polynit Heatseal Wipes in Refillable Tub, Contec®
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PROSAT®
PSPN0010
33500-466CS 367.32 USD InStock
33500-466
PROSAT® Polynit Heatseal Wipes in Refillable Tub, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Polyester wipes are chemically resistant and exceptionally low in particles and fibers. An interlock knit creates a durable fabric, the addition of a periodic additional stitch of a no-run interlock knit prevents the fabric unravelling creating an even more durable fabric with strong edges.
  • Recommended for ISO Class 3–5 environments
  • Presaturated with 9% semi conductor grade isopropanol and 91% DI water
  • Ideal for microelectronic applications
  • Sealed edge to reduce particle and fiber generation
  • 100% knitted polyester is abrasion and chemical resistant
  • Low in both particles and extractable contaminants
  • Presaturated wipes reduces VOC emissions and solvent use
The wipes are laundered and packed in an ISO Class 4 cleanroom. Through years of continuous improvement, the process minimizes non-volatile residues (NVRs) and particles, yet result in a sorbent wipe with excellent functionality.
Presaturated wipes ensure consistent saturation of each wipe independent of operator. Presaturated wipes can increase solvent control and accountability as well as reduce VOC emissions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty