Would you like to log in again?
Sontara® AC™ Aircraft Wipers, Contec®
Supplier: Contec
Sontara® AC™
AC12165 AC1212 AC1213 AC9165
33500-496CS 116.64 USD InStock
33500-496 76238-418 89012-918 33500-494
Sontara® AC™ Aircraft Wipers, Contec®
Wipes Disposable Wipes and Towels Wipes for General Applications
Sontara® AC™ aircraft wipes leave surfaces clean because they are extremely low-linting, and they tackle tough cleaning agents without falling apart.
  • Low-lint construction is suitable for aircraft manufacturing and maintenance
  • Durable fabric for tough cleaning applications
These are specially engineered to resist snagging on metal parts and corners. That's why they are superior to paper wipes, which snag and tear, leaving lint behind. These can also outperform cheesecloth, which can leave strings behind when it snags on a rivet. Their strength and durability make them ideal for a wide variety of general-purpose and pre-paint applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty