Would you like to log in again?
ECO Containers for ECO Funnels, CP Lab Safety
Supplier: CP Lab Safety
EC12751 EC-20060-IP-DHB EC-2009 EC-20010 EC-2004 EC-20020 EC-2002 EC12754 EC-20060-IC-DHB EC-20020DRN
33500-982EA 533.4 USD InStock
33500-982 33501-880 33501-882 33501-884 33501-874 33500-972 33500-978 33501-876 33501-878 33501-868
ECO Containers for ECO Funnels, CP Lab Safety
Sample Containers General Purpose Containers
ECO Funnels® are designed to fit a wide variety of standard Nalgene® carboys, Justrite® poly waste cans and other containers. The selection below includes the most common containers used with the ECO Funnel laboratory.
  • Made from PE
  • Good chemical resistance when glass is not an option
Containers with 38 mm, 53 mm, and 70 mm openings will typically work with one of the ECO Funnels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty