Would you like to log in again?
Digital Temperature Controller with Thermocouple, Chemglass
Supplier: Chemglass
CG-15001-30
33501-374EA 1753.85 USD InStock
33501-374
Digital Temperature Controller with Thermocouple, Chemglass
Controllers Temperature Controllers
PID controller for use with CG-1100 oil baths
  • Controller has 1500 watts of power and will control temperatures to ± 1.0°C
Ordering information: Supplied complete with CG-15001-25 Type "J" Thermocouple.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty