Would you like to log in again?
Polynit Heatseal Wipes, Contec®
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PN-99 IR PNHS-1620B PNHS-1212B PNHS-66 PNHS-44B LWPS0027 PNHS-99B PNHS-22B PNHS-1212 PNHS-44 PNHS-99B/150 PNHS-99 LWPS0006 LWPS0007
33501-474CS 1055.39 USD InStock
33501-474 68311-010 68311-008 68310-183 68310-194 89128-802 68310-180 68310-192 68300-556 76289-764 89217-354 10119-924 68310-176 62111-169
Polynit Heatseal Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polynit Heatseal Wipes are made of 100% knitted polyester that is laser cut to bond the fibers at the edges of the wipe. This creates a very clean, non-abrasive edge that allows full utilization of the wipe surface.
  • Recommended for ISO Class 3–5 environments for nonsterile wipes
  • Recommended for Grade A/B or ISO Class 5 environments for sterile wipes
  • Laundered knitted 100% polyester fabric for extremely low levels of particles and fibers
  • Heat sealed edges help prevent particle contamination
  • 100% knitted polyester is abrasion and chemical resistant
  • Laser-cut, sealed edge prevents release of fibers
Polynit Heatseal Wipes are exceptionally low in particles and extractable residue, making them ideal for critical cleaning. The wipes are laundered and packaged in an ISO Class 4 cleanroom. They have good sorbency with solvents.
The sterile wipes are gamma irradiated and validated per AAMI guidelines to a 10⁻⁶ SAL. They can be used in the highest grade life science cleanrooms.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty