Would you like to log in again?
Circular Reaction Blocks, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1991-01 CG-1991-05 CG-1991-02 CG-1991-04 CG-1991-03 CG-1991-06
33501-530EA 538.52 USD InStock
33501-530 33501-532 33501-522 33501-524 33501-526 33501-528
Circular Reaction Blocks, Chemglass
Block Heaters Blocks
Round reaction blocks are designed to fit directly on 135 mm diameter, round top hot plate stirrers.
  • Excellent heat transfer
  • Holes spaced for optimum stirring of all vials
  • Color-anodized for durability and easy identification
Aluminum construction offers excellent heat transfer without interfering with magnetic stirring. A thermowell is provided for insertion of a temperature sensor or contact thermometer.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty