Would you like to log in again?
COUPLING TRUSTIR FLEXSHFT 10MM
Supplier: Chemglass
Tru-Stir* Stirrer Shaft Coupling^ Fits 10mm stirrer shaft O.D., Upper coupling 7.9mm Male, For use with Flexible Shaft.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COUPLING TRUSTIR FLEXSHFT 10MM 33501-640 CG-2044-04 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty