Would you like to log in again?
Adapters, Rotary Evaporator Bump Trap, Fritted, R200 and R205, Chemglass
Supplier: Chemglass
CG-1317-C-10
33501-712EA 307.38 USD InStock
33501-712
Adapters, Rotary Evaporator Bump Trap, Fritted, R200 and R205, Chemglass
Adapters Adapters, Joints with Tube Connectors
Vapor tube/bump trap adapter is designed to be used with the Büchi® R-200/205 rotary evaporators.
  • This adapter combines a vapor tube adapter and a bump trap
  • Trap has an extra coarse fritted disc sealed between the lower inner joint and the top flange
  • Adapters have a 50 mm OD top flange with a tapered interior to fit the Büchi® sealing ring
Lower portion of the adapter has the listed standard taper inner joint. Each adapter is supplied complete with the corresponding size Keck clip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty