Would you like to log in again?
Funnels for FLASH Chromatography, California Pacific Lab, CP Lab Safety
Supplier: CP Lab Safety
CF-240 CF-60 CF-3005 CF-120 CF-960WN CF-960
33501-892EA 75.79 USD InStock
33501-892 33501-894 33501-896 33501-898 33501-900 47735-396
Funnels for FLASH Chromatography, California Pacific Lab, CP Lab Safety
Funnels Filtration Funnels Filter Funnels
Ideal for use in FLASH chromatography applications
Made of unbreakable polypropylene. Funnels feature a closed-end design that facilitates the gentle addition of a solvent to the column. The funnel stem is perforated to permit solvent to be poured down the solvent wall. The insertion of the funnel into the column will not disturb the top layer of sand or silica gel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty