Would you like to log in again?
Flasks, Heavy Wall, Apollo, Single Neck, Chemglass
Supplier: Chemglass
CG-1512-A-01 CG-1512-A-04 CG-1512-A-03 CG-1512-A-05 CG-1512-A-02 CG-1512-A-06
33502-618EA 110.76 USD InStock
33502-618 89028-582 89028-590 89028-586 89028-584 89028-588
Flasks, Heavy Wall, Apollo, Single Neck, Chemglass
Flasks Flat Bottom Flasks
Heavy wall single neck flask with a standard taper outer joint.
  • High-quality heavy wall flasks are short neck
  • Flasks feature a low profile design
  • Which allows the flask to sit upright on the bench
  • Permits easy access of products with a spatula or policeman
The flask contour is excellent for controlled evaporation of foaming solvents during rotary evaporation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty