Would you like to log in again?
Vacuum Filtration Adapters, with PTFE Stopcocks, Chemglass
Supplier: Chemglass
CG-1053-A-01 CG-1053-A-03
33502-688EA 244.13 USD InStock
33502-688 33502-684
Vacuum Filtration Adapters, with PTFE Stopcocks, Chemglass
Adapters Adapters, Conical Ground Joints
Adapter used for reduced pressure filtrations using plain stem buchner funnels.
  • Equipped with PTFE stopcock
  • Top of adapter is tooled to accept the listed size of Pluro stopper
Stopcock permits control of vacuum source.
Delivery information: Adapters are not supplied with stoppers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty