Would you like to log in again?
CONSTIX®, Cotton Swabs, SC-4 Series, Contec®
Supplier: Contec
CONSTIX®
SC-4
33502-698CS 496.22 USD InStock
33502-698
CONSTIX®, Cotton Swabs, SC-4 Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SC-4 cotton swabs have double-ended hard, sharp points. SC-4 swabs consist of high-quality, long staple-length cotton, tightly wrapped and shaped on both ends of high strength, wound paper handles. The heads are held to the handles using a methyl-cellulose binder.
  • High-quality cotton fibers provide excellent particle entrapment and retention
  • Methyl-cellulose binder minimizes fiber shedding
The high-quality cotton fibers provide exceptional sorbency of most solvents and solutions. The heads are very tightly wrapped and shaped using a methyl-cellulose binder which minimizes fiber shedding while maintaining low levels of extractable residue. The SC series cotton swabs are absorbent to most common solvents including isopropyl alcohol and acetone, and are ideal for use with aqueous based solutions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty